Learn More
tert-Butylamine, 99%
CAS: 75-64-9 | C4H11N | 73.13 g/mol
Supplier: Thermo Scientific Chemicals 107820010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| tert-Butylamine | |
| -67.0°C | |
| 46.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.3770 to 1.3790 | |
| MFCD00008050 | |
| 04,173 | |
| 0.69 | |
| tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert | |
| YBRBMKDOPFTVDT-UHFFFAOYSA-N | |
| 2-methylpropan-2-amine | |
| 6385 | |
| CHEBI:44639 | |
| 99% |
| 75-64-9 | |
| 0.6900g/mL | |
| −38°C | |
| 98.5% min. (GC) | |
| C4H11N | |
| (CH3)3CNH2 | |
| 1 L | |
| 01,84; 02,43; 10,62; 11,138 | |
| 15,1547 | |
| Solubility in water: 1000g/L (25°C). Other solubilities: miscible with alcohol,soluble in common organic solvents | |
| CC(C)(C)N | |
| 73.13 | |
| 0.5 mPa.s (20°C) | |
| 73.13 | |
| Liquid |
Chemical Identifiers
| 75-64-9 | |
| 73.13 | |
| YBRBMKDOPFTVDT-UHFFFAOYSA-N | |
| 6385 | |
| 2-methylpropan-2-amine |
| C4H11N | |
| MFCD00008050 | |
| tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert | |
| CHEBI:44639 | |
| CC(C)(C)N |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 200-888-1
RUO – Research Use Only