missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylamine, 99%, Thermo Scientific Chemicals

$41.38 - $159.10

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC107820050
SDS View Documents
Thermo Scientific Chemicals
107820050
5 mL Glass Bottle
Each for $41.38
 
AC107820010
SDS View Documents
Thermo Scientific Chemicals
107820010
1 L Glass Bottle
Each for $159.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-64-9
73.13
YBRBMKDOPFTVDT-UHFFFAOYSA-N
6385
2-methylpropan-2-amine
C4H11N
MFCD00008050
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
CHEBI:44639
CC(C)(C)N

Specifications

75-64-9
0.6900g/mL
−38°C
98.5% min. (GC)
C4H11N
(CH3)3CNH2
5 mL
01,84; 02,43; 10,62; 11,138
15,1547
Solubility in water: 1000g/L (25°C). Other solubilities: miscible with alcohol,soluble in common organic solvents
CC(C)(C)N
73.13
0.5 mPa.s (20°C)
73.13
Liquid
-67.0°C
46.0°C
Authentic
Glass Bottle
1.3770 to 1.3790
MFCD00008050
04,173
0.69
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
YBRBMKDOPFTVDT-UHFFFAOYSA-N
2-methylpropan-2-amine
6385
CHEBI:44639
99%
tert-Butylamine
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-888-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only