missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 396541000

 View more versions of this product

Catalog No. AC396541000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyllithium
Colorless to Yellow
36.0°C to 38.0°C
AcroSeal™ Glass bottle
(CH3)3CLi
100 mL
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
[Li]C(C)(C)C
64.06
64.04
Solution
594-19-4,109-66-0
0.6900g/mL
−56°C
C4H9Li
MFCD00008795
0.69
BKDLGMUIXWPYGD-UHFFFAOYSA-N
lithium;2-methylpropane
638178
17 to 19% (as BuLi)

Chemical Identifiers

594-19-4
64.06
BKDLGMUIXWPYGD-UHFFFAOYSA-N
638178
C4H9Li
MFCD00008795
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
[Li]C(C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from any possible contact with wa

GHS Signal Word: Danger

EINECSNumber : 209-831-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only