missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals

$210.81 - $538.44

Chemical Identifiers

CAS 594-19-4
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00008795
InChI Key BKDLGMUIXWPYGD-UHFFFAOYSA-N
Synonym tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
PubChem CID 638178
SMILES [Li]C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC396541000
SDS View Documents
Thermo Scientific Chemicals
396541000
100 mL AcroSeal™ Glass bottle
Each for $210.81
Only null left
 
AC396548000
SDS View Documents
Thermo Scientific Chemicals
396548000
800 mL AcroSeal™ Glass bottle
Each for $538.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

594-19-4
64.06
BKDLGMUIXWPYGD-UHFFFAOYSA-N
638178
C4H9Li
MFCD00008795
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
[Li]C(C)(C)C

Specifications

594-19-4,109-66-0
0.6900g/mL
−56°C
C4H9Li
MFCD00008795
0.69
BKDLGMUIXWPYGD-UHFFFAOYSA-N
lithium;2-methylpropane
638178
17 to 19% (as BuLi)
tert-Butyllithium
Colorless to Yellow
36.0°C to 38.0°C
AcroSeal™ Glass bottle
(CH3)3CLi
100 mL
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
[Li]C(C)(C)C
64.06
64.04
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from any possible contact with wa

GHS Signal Word: Danger

EINECSNumber : 209-831-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only