Terumo™ Tuberculin Syringes
Tuberculin Luer Slip syringes in a tamper-evident package
Supplier:  Terumo™ SS01T2713
 View more versions of this product
Catalog No. 22-293148
Explore available promotions
Description
Description
  • Syringes are equipped with clear, bold scale markings to assure dosage control and accuracy
  • Tamper-evident packaging for greater protection of the sterile contents
Specifications
Specifications
27 G
13 mm
Removable
0.5 in.
1 mL