Learn More
Tetra-n-butylammonium iodide, 98%, Thermo Scientific Chemicals
$66.33 - $654.59
Chemical Identifiers
CAS | 311-28-4 |
---|---|
Molecular Formula | C16H36IN |
Molecular Weight (g/mol) | 369.38 |
MDL Number | MFCD00011636 |
InChI Key | DPKBAXPHAYBPRL-UHFFFAOYSA-M |
Synonym | tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide |
PubChem CID | 67553 |
IUPAC Name | tetrabutylazanium;iodide |
SMILES | [I-].CCCC[N+](CCCC)(CCCC)CCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1548418
|
Thermo Scientific Chemicals
A1548418 |
50 g |
Each for $66.33
|
|
|||||
AAA1548430
|
Thermo Scientific Chemicals
A1548430 |
250 g |
Each for $238.62
|
|
|||||
AAA154840B
|
Thermo Scientific Chemicals
A154840B |
1000 g |
Each for $654.59
|
|
|||||
Description
Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.
Solubility
Soluble in water and methanol. Insoluble in benzene.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
311-28-4 | |
369.38 | |
DPKBAXPHAYBPRL-UHFFFAOYSA-M | |
67553 | |
[I-].CCCC[N+](CCCC)(CCCC)CCCC |
C16H36IN | |
MFCD00011636 | |
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide | |
tetrabutylazanium;iodide |
Specifications
311-28-4 | |
C16H36IN | |
50 g | |
Hygroscopic; Light sensitive | |
Soluble in water and methanol. Insoluble in benzene. | |
[I-].CCCC[N+](CCCC)(CCCC)CCCC | |
369.38 | |
369.38 | |
Tetra-n-butylammonium iodide |
142°C to 148°C | |
MFCD00011636 | |
3916152 | |
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide | |
DPKBAXPHAYBPRL-UHFFFAOYSA-M | |
tetrabutylazanium;iodide | |
67553 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 206-220-5
RTECSNumber : BS5450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only