missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 185685000

 View more versions of this product

Catalog No. AC185685000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium bromide
1643-19-2
Cream to White
99% min. (Argentometry)
C16H36BrN
MFCD00011633
04,477; 05,644; 07,353; 17,363
Solubility in water: 600g/L (20°C).
CCCC[N+](CCCC)(CCCC)CCCC.[Br-]
322.36
CHEBI:51993
99+%
99+%
100.0°C to 104.0°C
Authentic
Glass bottle
[CH3(CH2)3]4NBr
500 g
tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
JRMUNVKIHCOMHV-UHFFFAOYSA-M
tetrabutylazanium;bromide
74236
322.36
Crystalline Powder

Chemical Identifiers

1643-19-2
322.36
JRMUNVKIHCOMHV-UHFFFAOYSA-M
74236
tetrabutylazanium;bromide
C16H36BrN
MFCD00011633
tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
CHEBI:51993
CCCC[N+](CCCC)(CCCC)CCCC.[Br-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-699-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only