Learn More
Tetrabutylammonium bromide, 99+%
CAS: 1643-19-2 | C16H36BrN | 322.36 g/mol
$97.01 - $469.22
Chemical Identifiers
| CAS | 1643-19-2 |
|---|---|
| Molecular Formula | C16H36BrN |
| Molecular Weight (g/mol) | 322.36 |
| MDL Number | MFCD00011633 |
| InChI Key | JRMUNVKIHCOMHV-UHFFFAOYSA-M |
| Synonym | tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r |
| PubChem CID | 74236 |
| ChEBI | CHEBI:51993 |
| IUPAC Name | tetrabutylazanium;bromide |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC185680250
|
Thermo Scientific Chemicals
185680250 |
25 g | Glass bottle |
Each for $97.01
|
|
||||
|
AC185680500
|
Thermo Scientific Chemicals
185680500 |
50 g | Glass bottle |
Each for $110.04
|
|
||||
|
AC185681000
|
Thermo Scientific Chemicals
185681000 |
100 g | Glass bottle |
Each for $155.65
|
|
||||
|
AC185685000
|
Thermo Scientific Chemicals
185685000 |
500 g | Glass bottle |
Each for $469.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
| 1643-19-2 | |
| 322.36 | |
| JRMUNVKIHCOMHV-UHFFFAOYSA-M | |
| 74236 | |
| tetrabutylazanium;bromide |
| C16H36BrN | |
| MFCD00011633 | |
| tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r | |
| CHEBI:51993 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[Br-] |
Specifications
| 1643-19-2 | |
| Cream to White | |
| 99% min. (Argentometry) | |
| C16H36BrN | |
| MFCD00011633 | |
| 04,477; 05,644; 07,353; 17,363 | |
| Solubility in water: 600g/L (20°C). | |
| CCCC[N+](CCCC)(CCCC)CCCC.[Br-] | |
| 322.36 | |
| CHEBI:51993 | |
| 99+% | |
| Tetrabutylammonium bromide, 99+% |
| 100.0°C to 104.0°C | |
| Authentic | |
| Glass bottle | |
| [CH3(CH2)3]4NBr | |
| 25 g | |
| tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r | |
| JRMUNVKIHCOMHV-UHFFFAOYSA-M | |
| tetrabutylazanium;bromide | |
| 74236 | |
| 322.36 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-699-2
TSCA : TSCA
RUO – Research Use Only