missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 434830250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium hydroxide 30-hydrate | |
White | |
Crystals | |
95+% | |
MFCD00149573 | |
DFGIRVKFXSRUOX-UHFFFAOYSA-M | |
tetrabutylazanium;hydroxide;triacontahydrate | |
16218633 | |
≥95% |
27.0°C to 30.0°C | |
147741-30-8 | |
25 g | |
C16H97NO31 | |
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30 | |
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
799.93 | |
799.93 | |
Glass bottle |
Chemical Identifiers
147741-30-8 | |
799.93 | |
DFGIRVKFXSRUOX-UHFFFAOYSA-M | |
16218633 |
C16H97NO31 | |
MFCD00149573 | |
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30 | |
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
RUO – Research Use Only