missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

$168.09 - $415.46

Chemical Identifiers

CAS 147741-30-8
Molecular Formula C16H97NO31
Molecular Weight (g/mol) 799.93
MDL Number MFCD00149573
InChI Key DFGIRVKFXSRUOX-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
PubChem CID 16218633
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434830250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
434830250
25 g Glass bottle
Each for $168.09
 
AC434831000
SDS View Documents
Thermo Scientific Chemicals
434831000
100 g Glass bottle
Each for $415.46
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

147741-30-8
799.93
DFGIRVKFXSRUOX-UHFFFAOYSA-M
16218633
C16H97NO31
MFCD00149573
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

27.0°C to 30.0°C
147741-30-8
25 g
C16H97NO31
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
799.93
799.93
Glass bottle
White
Crystals
95+%
MFCD00149573
DFGIRVKFXSRUOX-UHFFFAOYSA-M
tetrabutylazanium;hydroxide;triacontahydrate
16218633
≥95%
Tetrabutylammonium hydroxide 30-hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only