missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 434831000

 View more versions of this product

Catalog No. AC434831000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium hydroxide 30-hydrate
White
Crystals
95+%
MFCD00149573
DFGIRVKFXSRUOX-UHFFFAOYSA-M
tetrabutylazanium triacontahydrate hydroxide
16218633
≥95%
27.0°C to 30.0°C
147741-30-8
100 g
C16H97NO31
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
799.93
799.93
Glass bottle

Chemical Identifiers

147741-30-8
799.93
DFGIRVKFXSRUOX-UHFFFAOYSA-M
16218633
C16H97NO31
MFCD00149573
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only