Learn More
Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 353090250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium tetrafluoroborate | |
429-42-5 | |
White | |
97.5% min | |
C16H36BF4N | |
25 g | |
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4; | |
NNZZSJSQYOFZAM-UHFFFAOYSA-N | |
tetrabutylazanium;tetrafluoroborate | |
67932 | |
98% |
95% | |
156°C to 163°C | |
Authentic | |
Plastic bottle | |
MFCD00011634 | |
04, III, 293 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,1g/10 mL methanol | |
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC | |
329.27 | |
329.28 | |
Crystalline Powder |
Chemical Identifiers
429-42-5 | |
329.27 | |
NNZZSJSQYOFZAM-UHFFFAOYSA-N | |
67932 | |
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC |
C16H36BF4N | |
MFCD00011634 | |
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4; | |
tetrabutylazanium;tetrafluoroborate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 207-058-8
TSCA : TSCA
RUO – Research Use Only