missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

$202.33 - $522.25

Chemical Identifiers

CAS 429-42-5
Molecular Formula C16H36BF4N
Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
PubChem CID 67932
IUPAC Name tetrabutylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC353090250
SDS View Documents
Thermo Scientific Chemicals
353090250
25 g Plastic bottle
Each for $202.33
 
AC353091000
SDS View Documents
Thermo Scientific Chemicals
353091000
100 g Plastic bottle
Each for $522.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

429-42-5
329.27
NNZZSJSQYOFZAM-UHFFFAOYSA-N
67932
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
C16H36BF4N
MFCD00011634
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
tetrabutylazanium;tetrafluoroborate

Specifications

429-42-5
White
97.5% min
C16H36BF4N
25 g
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylazanium;tetrafluoroborate
67932
98%
Tetrabutylammonium tetrafluoroborate, 95%
156°C to 163°C
Authentic
Plastic bottle
MFCD00011634
04, III, 293
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,1g/10 mL methanol
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
329.27
329.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 207-058-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only