missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 353091000

 View more versions of this product

Catalog No. AC353091000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium tetrafluoroborate
429-42-5
White
97.5% min
C16H36BF4N
100 g
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylazanium;tetrafluoroborate
67932
98%
95%
156°C to 163°C
Authentic
Plastic bottle
MFCD00011634
04, III, 293
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,1g/10 mL methanol
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
329.27
329.28
Crystalline Powder

Chemical Identifiers

429-42-5
329.27
NNZZSJSQYOFZAM-UHFFFAOYSA-N
67932
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
C16H36BF4N
MFCD00011634
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
tetrabutylazanium;tetrafluoroborate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 207-058-8

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only