Learn More
Tetraethylene glycol, 99%, Thermo Scientific Chemicals
$57.12 - $189.10
Chemical Identifiers
CAS | 112-60-7 |
---|---|
Molecular Formula | C8H18O5 |
Molecular Weight (g/mol) | 194.227 |
MDL Number | MFCD00002879 |
InChI Key | UWHCKJMYHZGTIT-UHFFFAOYSA-N |
Synonym | tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843 |
PubChem CID | 8200 |
ChEBI | CHEBI:44920 |
IUPAC Name | 2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol |
SMILES | C(COCCOCCOCCO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB23880AP
|
Thermo Scientific Chemicals
B23880AP |
500 mL |
Each for $57.12
|
|
|||||
AAB238800F
|
Thermo Scientific Chemicals
B238800F |
2500 mL |
Each for $189.10
|
|
|||||
Description
Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.
Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerin, pyridine, aldehydes, carbon tetrachloride and diethyl ether. Immiscible with oil, benzene, toluene, fat and aliphatic hydrocarbons.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
112-60-7 | |
194.227 | |
UWHCKJMYHZGTIT-UHFFFAOYSA-N | |
8200 | |
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol |
C8H18O5 | |
MFCD00002879 | |
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843 | |
CHEBI:44920 | |
C(COCCOCCOCCO)O |
Specifications
112-60-7 | |
1.125 | |
313°C to 314°C | |
Odorless | |
1.459 | |
500 mL | |
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843 | |
UWHCKJMYHZGTIT-UHFFFAOYSA-N | |
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol | |
8200 | |
194.23 | |
Tetraethylene glycol |
-4°C | |
8.5 | |
193°C (379°F) | |
C8H18O5 | |
MFCD00002879 | |
1634320 | |
Miscible with water,ethanol,acetone,acetic acid,glycerin,pyridine,aldehydes,carbon tetrachloride and diethyl ether. Immiscible with oil,benzene,toluene,fat and aliphatic hydrocarbons. | |
C(COCCOCCOCCO)O | |
194.227 | |
CHEBI:44920 | |
99% |
Safety and Handling
P201-P202-P262-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P314-P332+P313-P362-P501c
H315-H320-H341-H360-H373
EINECSNumber : 203-989-9
RTECSNumber : XC2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only