missing translation for 'onlineSavingsMsg'
Learn More

tetrahydro-2h-pyran-4-ylmethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC29909DE

 View more versions of this product

Catalog No. CC29909DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Tetrahydro-2H-pyran-4-ylmethanol
97%
C6H12O2
5g
YSNVSVCWTBLLRW-UHFFFAOYSA-N
oxan-4-ylmethanol
2773573
97%
14774-37-9
Amber Glass Bottle
MFCD00457804
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
OCC1CCOCC1
116.16
116.16

Chemical Identifiers

14774-37-9
116.16
YSNVSVCWTBLLRW-UHFFFAOYSA-N
2773573
OCC1CCOCC1
C6H12O2
MFCD00457804
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
oxan-4-ylmethanol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions