missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tetrahydro-2h-pyran-4-ylmethanol, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 14774-37-9 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00457804 |
InChI Key | YSNVSVCWTBLLRW-UHFFFAOYSA-N |
Synonym | tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol |
PubChem CID | 2773573 |
IUPAC Name | oxan-4-ylmethanol |
SMILES | OCC1CCOCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC29909CB
|
Thermo Scientific Chemicals
CC29909CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC29909DA
|
Thermo Scientific Chemicals
CC29909DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC29909DE
|
Thermo Scientific Chemicals
CC29909DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
14774-37-9 | |
116.16 | |
YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
2773573 | |
OCC1CCOCC1 |
C6H12O2 | |
MFCD00457804 | |
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
oxan-4-ylmethanol |
Specifications
14774-37-9 | |
Amber Glass Bottle | |
MFCD00457804 | |
tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol | |
OCC1CCOCC1 | |
116.16 | |
116.16 | |
Tetrahydro-2H-pyran-4-ylmethanol |
97% | |
C6H12O2 | |
250mg | |
YSNVSVCWTBLLRW-UHFFFAOYSA-N | |
oxan-4-ylmethanol | |
2773573 | |
97% |