missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrothiophen-3-one, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1100209

 View more versions of this product

Catalog No. AAA1100209


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrahydrothiophen-3-one
1.192
77°C (170°F)
C4H6OS
MFCD00005412
105201
DSXFPRKPFJRPIB-UHFFFAOYSA-N
thiolan-3-one
61252
97%
1003-04-9
174°C to 175°C
Characteristic
1.529
10 g
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
O=C1CCSC1
102.15
102.16

Chemical Identifiers

1003-04-9
102.15
DSXFPRKPFJRPIB-UHFFFAOYSA-N
61252
O=C1CCSC1
C4H6OS
MFCD00005412
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
thiolan-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 213-698-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only