Learn More
Tetrahydrothiophen-3-one, 97%, Thermo Scientific Chemicals
$53.33 - $161.25
Chemical Identifiers
CAS | 1003-04-9 |
---|---|
Molecular Formula | C4H6OS |
Molecular Weight (g/mol) | 102.15 |
MDL Number | MFCD00005412 |
InChI Key | DSXFPRKPFJRPIB-UHFFFAOYSA-N |
Synonym | tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene |
PubChem CID | 61252 |
IUPAC Name | thiolan-3-one |
SMILES | O=C1CCSC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1100206
|
Thermo Scientific Chemicals
A1100206 |
5 g |
Each for $53.33
|
|
|||||
AAA1100209
|
Thermo Scientific Chemicals
A1100209 |
10 g |
Each for $86.21
|
|
|||||
AAA1100214
|
Thermo Scientific Chemicals
A1100214 |
25 g |
Each for $161.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1003-04-9 | |
102.15 | |
DSXFPRKPFJRPIB-UHFFFAOYSA-N | |
61252 | |
O=C1CCSC1 |
C4H6OS | |
MFCD00005412 | |
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene | |
thiolan-3-one |
Specifications
1003-04-9 | |
174°C to 175°C | |
Characteristic | |
1.529 | |
5 g | |
tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene | |
O=C1CCSC1 | |
102.15 | |
102.16 | |
Tetrahydrothiophen-3-one |
1.192 | |
77°C (170°F) | |
C4H6OS | |
MFCD00005412 | |
105201 | |
DSXFPRKPFJRPIB-UHFFFAOYSA-N | |
thiolan-3-one | |
61252 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 213-698-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only