missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrothiopyran-4-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC61963DA

 View more versions of this product

Catalog No. CC61963DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Tetrahydrothiopyran-4-ylmethylamine hydrochloride
96.5
97%
C6H14ClNS
1g
DREBXEFIPXBLAP-UHFFFAOYSA-N
thian-4-ylmethanamine;hydrochloride
24229644
97%
950603-21-1
100.0
Amber Glass Bottle
MFCD09879925
tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
C1CSCCC1CN.Cl
167.695
167.7

Chemical Identifiers

950603-21-1
167.695
DREBXEFIPXBLAP-UHFFFAOYSA-N
24229644
C1CSCCC1CN.Cl
C6H14ClNS
MFCD09879925
tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
thian-4-ylmethanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions