missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydrothiopyran-4-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 950603-21-1
Molecular Formula C6H14ClNS
Molecular Weight (g/mol) 167.695
MDL Number MFCD09879925
InChI Key DREBXEFIPXBLAP-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
PubChem CID 24229644
IUPAC Name thian-4-ylmethanamine;hydrochloride
SMILES C1CSCCC1CN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC61963CB
SDS View Documents
Thermo Scientific Chemicals
CC61963CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61963DA
SDS View Documents
Thermo Scientific Chemicals
CC61963DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

950603-21-1
167.695
DREBXEFIPXBLAP-UHFFFAOYSA-N
24229644
C1CSCCC1CN.Cl
C6H14ClNS
MFCD09879925
tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
thian-4-ylmethanamine;hydrochloride

Specifications

950603-21-1
100.0
Amber Glass Bottle
MFCD09879925
tetrahydro-2h-thiopyran-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethyl amine hydrochloride, thian-4-ylmethanamine hydrochloride, thian-4-ylmethylamine, chloride, 1-thian-4-yl methanamine hydrochloride, tetrahydrothiopyran-4-ylmethylamine hydrochloride, 1-thian-4-yl methanamine-hydrogen chloride 1/1, tetrahydro-2h-thiopyran-4-yl methylamine hydrochloride
C1CSCCC1CN.Cl
167.695
167.7
Tetrahydrothiopyran-4-ylmethylamine hydrochloride
96.5
97%
C6H14ClNS
250mg
DREBXEFIPXBLAP-UHFFFAOYSA-N
thian-4-ylmethanamine;hydrochloride
24229644
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'