missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(acetonitrile)copper(I) tetrafluoroborate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 398760010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetrakis(acetonitrile)copper(I) tetrafluoroborate | |
Glass bottle | |
MFCD09265110 | |
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate | |
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F | |
314.56 | |
314.56 |
15418-29-8 | |
C8H12BCuF4N4 | |
1 g | |
YZGSKMIIVMCEFE-UHFFFAOYSA-N | |
acetonitrile;copper(1+);tetrafluoroborate | |
10990673 |
Chemical Identifiers
15418-29-8 | |
314.56 | |
YZGSKMIIVMCEFE-UHFFFAOYSA-N | |
acetonitrile;copper(1+);tetrafluoroborate |
C8H12BCuF4N4 | |
MFCD09265110 | |
10990673 | |
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO – Research Use Only