missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(acetonitrile)copper(I) tetrafluoroborate, Thermo Scientific Chemicals

$52.61 - $134.73

Chemical Identifiers

CAS 15418-29-8
Molecular Formula C8H12BCuF4N4
Molecular Weight (g/mol) 314.56
MDL Number MFCD09265110
InChI Key YZGSKMIIVMCEFE-UHFFFAOYSA-N
PubChem CID 10990673
IUPAC Name acetonitrile;copper(1+);tetrafluoroborate
SMILES [Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398760010
SDS View Documents
Thermo Scientific Chemicals
398760010
1 g Glass bottle
Each for $52.61
 
AC398760050
SDS View Documents
Thermo Scientific Chemicals
398760050
5 g Glass bottle
Each for $134.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15418-29-8
314.56
YZGSKMIIVMCEFE-UHFFFAOYSA-N
acetonitrile;copper(1+);tetrafluoroborate
C8H12BCuF4N4
MFCD09265110
10990673
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F

Specifications

15418-29-8
C8H12BCuF4N4
1 g
YZGSKMIIVMCEFE-UHFFFAOYSA-N
acetonitrile;copper(1+);tetrafluoroborate
10990673
Tetrakis(acetonitrile)copper(I) tetrafluoroborate
Glass bottle
MFCD09265110
tetrakis acetonitrile copper i tetrafluoroborate, cu mecn 4 bf4, cu mecn 4bf4, copper i tetra acetonitrile tetrafluoroborate, tetra acetonitrile copper 1+ tetrafluoroborate
[Cu+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F
314.56
314.56
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only