missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 203821000

 View more versions of this product

Catalog No. AC203821000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetramethyl orthosilicate
-2.0°C
121.0°C to 122.0°C
Authentic
Glass bottle
1.3670 to 1.3700
MFCD00008341
01,287
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
LFQCEHFDDXELDD-UHFFFAOYSA-N
tetramethyl silicate
12682
152.22
Liquid
681-84-5
1.0300g/mL
26°C
98.5% min. (GC)
C4H12O4Si
Si(OCH3)4
100 g
1.03
Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol
CO[Si](OC)(OC)OC
152.22
0.7 mPa.s (20°C)
99%

Chemical Identifiers

681-84-5
152.22
LFQCEHFDDXELDD-UHFFFAOYSA-N
12682
CO[Si](OC)(OC)OC
C4H12O4Si
MFCD00008341
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
tetramethyl silicate
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 211-656-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only