missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals
$77.91 - $421.17
Chemical Identifiers
CAS | 681-84-5 |
---|---|
Molecular Formula | C4H12O4Si |
Molecular Weight (g/mol) | 152.22 |
MDL Number | MFCD00008341 |
InChI Key | LFQCEHFDDXELDD-UHFFFAOYSA-N |
Synonym | tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate |
PubChem CID | 12682 |
IUPAC Name | tetramethyl silicate |
SMILES | CO[Si](OC)(OC)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC203820250
|
Thermo Scientific Chemicals
203820250 |
25 g | Glass bottle |
Each for $77.91
|
|
||||
AC203821000
|
Thermo Scientific Chemicals
203821000 |
100 g | Glass bottle |
Each for $158.54
|
|
||||
AC203825000
|
Thermo Scientific Chemicals
203825000 |
500 g | Glass bottle |
Each for $421.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
681-84-5 | |
152.22 | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
12682 | |
CO[Si](OC)(OC)OC |
C4H12O4Si | |
MFCD00008341 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
tetramethyl silicate |
Specifications
681-84-5 | |
1.0300g/mL | |
26°C | |
98.5% min. (GC) | |
C4H12O4Si | |
Si(OCH3)4 | |
25 g | |
1.03 | |
Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol | |
CO[Si](OC)(OC)OC | |
152.22 | |
0.7 mPa.s (20°C) | |
99% | |
Tetramethyl orthosilicate |
-2.0°C | |
121.0°C to 122.0°C | |
Authentic | |
Glass bottle | |
1.3670 to 1.3700 | |
MFCD00008341 | |
01,287 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
tetramethyl silicate | |
12682 | |
152.22 | |
Liquid |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 211-656-4
RUO – Research Use Only