missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 203825000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetramethyl orthosilicate | |
-2.0°C | |
121.0°C to 122.0°C | |
Authentic | |
Glass bottle | |
1.3670 to 1.3700 | |
MFCD00008341 | |
01,287 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
tetramethyl silicate | |
12682 | |
152.22 | |
Liquid |
681-84-5 | |
1.0300g/mL | |
26°C | |
98.5% min. (GC) | |
C4H12O4Si | |
Si(OCH3)4 | |
500 g | |
1.03 | |
Solubility in water: hydrolysis in water. Other solubilities: soluble in alcohol | |
CO[Si](OC)(OC)OC | |
152.22 | |
0.7 mPa.s (20°C) | |
99% |
Chemical Identifiers
681-84-5 | |
152.22 | |
LFQCEHFDDXELDD-UHFFFAOYSA-N | |
12682 | |
CO[Si](OC)(OC)OC |
C4H12O4Si | |
MFCD00008341 | |
tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate | |
tetramethyl silicate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 211-656-4
RUO – Research Use Only