missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1746714

 View more versions of this product

Catalog No. AAA1746714


Explore available promotions

Description

Description

It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications

Specifications

Tetramethylammonium hexafluorophosphate
>300°C
C4H12F6NP
MFCD00012012
UN2923
Moisture sensitive
Soluble in water.
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
219.111
219.1
558-32-7
Odorless
(CH3)4NPF6
25 g
3730394
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylazanium;hexafluorophosphate
11209
≥99%

Chemical Identifiers

558-32-7
219.111
ZFKULDQWLXAKIM-UHFFFAOYSA-N
11209
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
C4H12F6NP
MFCD00012012
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
tetramethylazanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 209-194-3

RTECSNumber : BS8000000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only