Learn More
Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific Chemicals
$260.96 - $727.73
Chemical Identifiers
CAS | 558-32-7 |
---|---|
Molecular Formula | C4H12F6NP |
Molecular Weight (g/mol) | 219.111 |
MDL Number | MFCD00012012 |
InChI Key | ZFKULDQWLXAKIM-UHFFFAOYSA-N |
Synonym | tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci |
PubChem CID | 11209 |
IUPAC Name | tetramethylazanium;hexafluorophosphate |
SMILES | C[N+](C)(C)C.F[P-](F)(F)(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1746714
|
Thermo Scientific Chemicals
A1746714 |
25 g |
Each for $260.96
|
|
|||||
AAA1746722
|
Thermo Scientific Chemicals
A1746722 |
100 g |
Each for $727.73
|
|
|||||
Description
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the oxidation of cytochrome c in nonaqueous solvents.
Solubility
Soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Chemical Identifiers
558-32-7 | |
219.111 | |
ZFKULDQWLXAKIM-UHFFFAOYSA-N | |
11209 | |
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F |
C4H12F6NP | |
MFCD00012012 | |
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci | |
tetramethylazanium;hexafluorophosphate |
Specifications
558-32-7 | |
Odorless | |
(CH3)4NPF6 | |
25 g | |
3730394 | |
tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci | |
ZFKULDQWLXAKIM-UHFFFAOYSA-N | |
tetramethylazanium;hexafluorophosphate | |
11209 | |
≥99% |
>300°C | |
C4H12F6NP | |
MFCD00012012 | |
UN2923 | |
Moisture sensitive | |
Soluble in water. | |
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F | |
219.111 | |
219.1 | |
Tetramethylammonium hexafluorophosphate |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H332-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 209-194-3
RTECSNumber : BS8000000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only