missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals
$88.89 - $249.21
Chemical Identifiers
CAS | 75-76-3 |
---|---|
Molecular Formula | C4H12Si |
Molecular Weight (g/mol) | 88.23 |
MDL Number | MFCD00008274 |
InChI Key | CZDYPVPMEAXLPK-UHFFFAOYSA-N |
Synonym | silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 |
PubChem CID | 6396 |
ChEBI | CHEBI:85361 |
IUPAC Name | tetramethylsilane |
SMILES | C[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC445900250
|
Thermo Scientific Chemicals
445900250 |
25 g | Glass bottle |
Each for $88.89
|
|
||||
AC445901000
|
Thermo Scientific Chemicals
445901000 |
100 g | Glass bottle |
Each for $249.21
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-76-3 | |
88.23 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
6396 | |
tetramethylsilane |
C4H12Si | |
MFCD00008274 | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CHEBI:85361 | |
C[Si](C)(C)C |
Specifications
75-76-3 | |
Colorless | |
26°C to 28°C | |
Glass bottle | |
1.3585 | |
MFCD00008274 | |
0.64 | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
tetramethylsilane | |
6396 | |
88.22 | |
Liquid |
-99°C | |
0.6400g/mL | |
-27°C | |
C4H12Si | |
Si(CH3)4 | |
25 g | |
15,9377 | |
Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents | |
C[Si](C)(C)C | |
88.23 | |
CHEBI:85361 | |
ACS Reagent | |
Tetramethylsilane |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 200-899-1
RUO – Research Use Only