Learn More
Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 204030500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)Specifications
Tetraphenylphosphonium chloride | |
2001-45-8 | |
White to Beige | |
97.5% min. (Argentometry) | |
C24H20ClP | |
MFCD00011916 | |
16, II, 373 | |
Solubility in water: soluble. | |
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
374.85 | |
374.84 | |
Crystalline Powder |
99% | |
275.0°C to 280.0°C | |
Authentic | |
Glass bottle | |
(C6H5)4PCl | |
50 g | |
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co | |
WAGFXJQAIZNSEQ-UHFFFAOYSA-M | |
tetraphenylphosphanium;chloride | |
164911 | |
98% |
Chemical Identifiers
2001-45-8 | |
374.85 | |
WAGFXJQAIZNSEQ-UHFFFAOYSA-M | |
164911 | |
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20ClP | |
MFCD00011916 | |
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co | |
tetraphenylphosphanium;chloride |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 217-890-3
RUO – Research Use Only