missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

$186.82 - $516.07

Chemical Identifiers

CAS 2001-45-8
Molecular Formula C24H20ClP
Molecular Weight (g/mol) 374.85
MDL Number MFCD00011916
InChI Key WAGFXJQAIZNSEQ-UHFFFAOYSA-M
Synonym tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
PubChem CID 164911
IUPAC Name tetraphenylphosphanium;chloride
SMILES [Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC204030100
SDS View Documents
Thermo Scientific Chemicals
204030100
10 g Glass bottle
Each for $186.82
 
AC204030500
SDS View Documents
Thermo Scientific Chemicals
204030500
50 g Glass bottle
Each for $516.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

2001-45-8
374.85
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
164911
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C24H20ClP
MFCD00011916
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
tetraphenylphosphanium;chloride

Specifications

2001-45-8
White to Beige
97.5% min. (Argentometry)
C24H20ClP
MFCD00011916
16, II, 373
Solubility in water: soluble.
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
374.85
374.84
Crystalline Powder
275.0°C to 280.0°C
Authentic
Glass bottle
(C6H5)4PCl
10 g
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
tetraphenylphosphanium;chloride
164911
98%
Tetraphenylphosphonium chloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 217-890-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only