Texwipe™ TOC Cleaning Validation Kits
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Supplier:  Texwipe™ TX3340
 View more versions of this product
Catalog No. 19-130-3720
Explore available promotions
Description
Description
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
  • Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
  • Prewashed vials have one-piece cap and septa design for easy sampling and analysis
  • Polyethylene overcap protects septum surface from contamination until time of analysis
  • Sloped shoulder design minimizes headspace for reduced background TOC level
  • Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
Sampling Swab
  • Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
  • Double layered, double knit polyester head entraps contaminants in knit structure during sampling
  • Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
  • Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
  • 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Specifications
Specifications
Sampling 12 production areas
12 40 mL clear vials bonded septa cap, 24 Low TOC Alpha™ Sampling Swab, 12 blank vial labels
18/Cs.