Texwipe™ TOC Cleaning Validation Kits
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Supplier: Texwipe™ TX3342
Description
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
- Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
- Prewashed vials have one-piece cap and septa design for easy sampling and analysis
- Polyethylene overcap protects septum surface from contamination until time of analysis
- Sloped shoulder design minimizes headspace for reduced background TOC level
- Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
- Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
- Double layered, double knit polyester head entraps contaminants in knit structure during sampling
- Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
- Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
- 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Specifications
Sampling 72 production areas | |
72 40 mL clear vials bonded septa cap, 144 Low TOC Alpha™ Sampling Swab, 72 blank vial labels | |
1/Cs. |