missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

METTLER TOLEDO™ TGA/DSC

The TGA/DSC 3+ provides trustworthy results using a TGA balance from the worldwide leader in weighing technology with position-independent weighing, automatic internal calibration weights, a wide measurement range, the best minimum weight performance and the highest weighing accuracy and precision.

Supplier:  METTLER TOLEDO™ 30319254

 View more versions of this product

Catalog No. 01915194

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • METTLER TOLEDO ultra-micro balance – rely on the leader in balance technology
  • Very low minimum weight on 5-gram-balances – measure samples accurately and precisely
  • No hangdown wire – Our rigid horizontal sample arm eliminates decades of frustration in TGA operation
  • Unparalleled performance – sub-microgram resolution over the entire weighing range
  • Wide temperature range – analyze samples from ambient temperature to 1600 °C
  • DSC heat flow measurement – for simultaneous detection of thermal effects
  • Built-in gas flow control – investigate your samples in defined atmospheres
  • Automated TGA-FTIR, TGA-MS and TGA-GC/MS systems – perform accurate evolved gas analyses using FTIR and MS
  • Modular concept protects your investment – fits to your current and future needs
Note: This is a configurable instrument; additional METTLER TOLEDO accessories and software are required.  Please contact your local Fisher Scientific Sales Representative to discuss the best configuration for your application.

Specifications

Specifications

±0.3 K
Large Furnace TGA/DSC with SDTA Sensor
150 K/min
1100°C
SDTA sensor consists of a platinum support with thermocouple that measures the sample temperature
1 Year
24.8 in.
17.7 in.
88.2 lb.
50/60 Hz
IEC/EN61010-1:2001
≤900 μL
Includes a 6 place microgram balance (XP1) from the worldwide leader in weighing technology with position independent weighing
≤900 μL
Manual
24.8 x 21.3 x 17.7 in.
21.3 in.
24.8 in.
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions