missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thallium(I) selenide, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03629604

 View more versions of this product

Catalog No. AA3629604


Explore available promotions

Description

Description

Thallium(I) selenide is used in molten form as a semiconductor. Semiconducting thallium(I) selenide is used for IR detection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thallium(I) selenide
15572-25-5
2 g
MFCD00049603
14,9267
487.7
9.05 g/mL
∼340°C
Lump
(metals basis)
UN1707
Insoluble in water and acids.
99.9%

Chemical Identifiers

15572-25-5
MFCD00049603
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

EINECSNumber : 239-627-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only