missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thallium(I) selenide, 99.9% (metals basis), Thermo Scientific Chemicals

$184.96 - $609.63

Chemical Identifiers

CAS 15572-25-5
MDL Number MFCD00049603
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3629604
View Documents
Thermo Scientific Chemicals
03629604
2 g
Each for $184.96
 
AA3629609
View Documents
Thermo Scientific Chemicals
03629609
10 g
Each for $609.63
 
Description

Description

Thallium(I) selenide is used in molten form as a semiconductor. Semiconducting thallium(I) selenide is used for IR detection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15572-25-5
MFCD00049603

Specifications

∼340°C
Lump
MFCD00049603
14,9267
487.7
9.05 g/mL
(metals basis)
15572-25-5
2 g
UN1707
Insoluble in water and acids.
99.9%
Thallium(I) selenide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

EINECSNumber : 239-627-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only