Therapak™ Biological Substance Category B Ambient Shipping Systems
Include all the necessary marking labels in a convenient format for application at the shipping location
Supplier: Therapak™ 37804
Includes: Each system supplied with absorbent pouch or sheet, cushioning material to secure the inner packaging (except swab canister), biohazard and Biological Substances Category B labels, transport box, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), list of contents card, and Packing Instructions.
Description
- For transport of blood, urine and other biological substances
- Choice of pre-assembled systems to accommodate a variety of shipping needs
Specifications
6 Tubes | |
95kPa medium canister, Aqui-Pak absorbent pouch, cushioning material, biohazard and biological substance labels, contents card, transport box | |
Biological Substance Category B Ambient |