Therapak™ Biological Substance Category B Ambient Shipping Systems
Include all the necessary marking labels in a convenient format for application at the shipping location
$291.12 - $824.85
Specifications
Type | Biological Substance Category B Ambient |
---|
Includes: Each system supplied with absorbent pouch or sheet, cushioning material to secure the inner packaging (except swab canister), biohazard and Biological Substances Category B labels, transport box, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), list of contents card, and Packing Instructions.
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
22-130-025
|
Therapak™
37801 |
95kPa bag, Aqui-Pak absorbent pouch, cushioning material, biohazard and biological substance labels, list of contents card, transport box | 4 Tubes |
Case of 16 Each for $357.88
|
|
||||
22-130-027
|
Therapak™
37804 |
95kPa medium canister, Aqui-Pak absorbent pouch, cushioning material, biohazard and biological substance labels, contents card, transport box | 6 Tubes |
Case of 16 Each for $460.00
|
|
||||
22-130-026
|
Therapak™
37803 |
95kPa small canister, Aqui-Pak absorbent pouch, cushioning material, biohazard and biological substance labels, contents card, transport box; | 4 Tubes |
Case of 36 Each for $824.85
|
|
||||
22-130-052
|
Therapak™
37925 |
Large 95kPa bag; 3 Aqui-Pak absorbent pouch, cushioning material, biohazard, Biological Substance Category B label, list of contents card, and transport box | 18 Tubes |
Case of 6 Each for $291.12
|
|
||||
22-130-419
|
Therapak™
37802 |
95kPa Bag; TearTech tube shuttle, cushioning material, biohazard and Biological Substance Category B labels, list of contents card, transport box | 6 Tubes |
Case of 16 Each for $488.36
|
|
||||
22-130-420
|
Therapak™
37980 |
95kPa Bag, Insulated envelope, gel wrap; Aqui-Pak absorbent pouch, cushioning material, biohazard and Biological Substance Category B labels, transport box, list of contents card | 6 Tubes |
Case of 15 Each for $617.18
|
|
||||
Description
- For transport of blood, urine and other biological substances
- Choice of pre-assembled systems to accommodate a variety of shipping needs
Specifications
Biological Substance Category B Ambient |