Therapak™ Exempt Human Specimen Refrigerated and Frozen Shipping Systems
For diagnostic specimens that must be shipped refrigerated or frozen
Supplier:  Therapak™ TR56878
 View more versions of this product
Catalog No. 22-130-432
Explore available promotions
Description
Description
  • Refrigerated Shipping System is ideal for overnight shipments via common carriers; includes small insulated cooler; transport box; 8 oz. cold packs; specimen bag; absorbent material; Exempt Human Specimen and biohazard labels; and cushioning material
  • Frozen Shipping System designed for overnight shipments of frozen (dry ice) specimens by common carriers; includes small insulated cooler; transport box; specimen bag; absorbent material; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
  • Combination Shipping System for shipments of frozen and ambient specimens; includes small insulated cooler; insulated envelope; two biohazard bags; two Aqui-Pak™ pouches (each holds four tubes); ambient gel wrap; transport box; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
Specifications
Specifications
Small insulated cooler, envelope; 2 Biohazard bags; 2 Aqui-Pak pouches (holds four tubes); ambient gel wrap; transport box; EHS, biohazard, dry ice labels, cushioning
For overnight shipping