Therapak™ Exempt Human Specimen Refrigerated and Frozen Shipping Systems
For diagnostic specimens that must be shipped refrigerated or frozen
$197.48 - $237.73
Catalog Number | Mfr. No. | Includes | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | ||||||
22-130-431
|
Therapak™
56877 |
Small insulated cooler; transport box; specimen bag; absorbent material; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material |
Case of 3 Each for $237.73
|
|
||||||
22-130-430
|
Therapak™
TR56876 |
Includes small insulated cooler; transport box, 8 oz. cold packs, specimen bag, absorbent material; EHS and biohazard labels; Cushioning material |
Case of 3 Each for $229.79
|
|
||||||
22-130-432
|
Therapak™
TR56878 |
Small insulated cooler, envelope; 2 Biohazard bags; 2 Aqui-Pak pouches (holds four tubes); ambient gel wrap; transport box; EHS, biohazard, dry ice labels, cushioning |
Case of 2 Each for $197.48
|
|
||||||
Description
- Refrigerated Shipping System is ideal for overnight shipments via common carriers; includes small insulated cooler; transport box; 8 oz. cold packs; specimen bag; absorbent material; Exempt Human Specimen and biohazard labels; and cushioning material
- Frozen Shipping System designed for overnight shipments of frozen (dry ice) specimens by common carriers; includes small insulated cooler; transport box; specimen bag; absorbent material; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
- Combination Shipping System for shipments of frozen and ambient specimens; includes small insulated cooler; insulated envelope; two biohazard bags; two Aqui-Pak™ pouches (each holds four tubes); ambient gel wrap; transport box; Exempt Human Specimen, biohazard, and dry ice labels, and cushioning material
Specifications
For overnight shipping |