Therapak™ Single Specimen Collection Kits
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Supplier: Therapak™ TR40012
Description
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis. Each kit contains either a 180 mL beaker style collection cup with pour spout or a leak-resistant 90 mL sterile container with cap and integrity seal. The transport bag features two sealable, tamper evident pockets and comes with absorbent material. The individually bagged kits are available in various configurations.
Specifications
Sterile | |
90 mL (3 oz.) Sterile Urine Container with Temperature Strip | |
90 mL |
For drugs of abuse testing, on-site testing or remote site collection | |
3 oz. |