Therapak™ UN3373 Biological Substance Category B Labels
Universally recognized marking for shipping biological substances by air
Supplier:  Therapak™ 54752
 View more versions of this product
Catalog No. 22-130-067
Explore available promotions
Includes: 500 labels per roll
Description
Description
  • Convenient, easy-to-use roll format
  • IATA Packing Instruction 650 compliant
  • For use with Biological Substance, Category B specimens that do not meet the criteria for inclusion into the Infectious Substance Category A classification
  • Unlike Category A specimens, Category B specimens do not cause permanent disability, life-threatening illnesses or fatal disease to humans or animals when exposure occurs
Specifications
Specifications
DOT, IATA
IATA Biological Substance Category B Label
500/Pk.
8.8 x 10 cm
Label
For shipping biological substances by air
3.5 x 4 in.