Thermo Scientific™ GeneJET FFPE DNA Purification Kit
Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue.
Supplier:  Thermo Scientific™ K0882
 View more versions of this product
Catalog No. FERK0882
Explore available promotions
Description
Description
Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue. Up to 8 sections (10 μm thickness) of FFPE sample from various tissues can be used for genomic DNA extraction without overnight incubation. Elimination of toxic reagents commonly used for deparaffinization (i.e., xylene) allows a safe DNA purification procedure with high DNA yields and excellent quality. Each preparation recovers up to 8 μg of genomic DNA that can be eluted in volumes from 20 μL allowing for concentrated samples. Due to high decrosslinking efficiency high quality purified DNA shows reliable performance in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or NGS library preparation.
Highlights
High quality DNA—suitable for PCR and qPCR
Fast—procedure can be completed within 2 hours
Safe—non toxic solvents
Efficient—yields up to 8 μg of purified DNA
Applications
• PCR
• qPCR
• NGS library preparation
Includes
• Proteinase K Solution
• RNase A Solution
• Digestion Buffer
• Binding Buffer
• Wash Buffer I (conc.)
• Wash Buffer II (conc.)
• Elution Buffer
• GeneJET DNA Purification Columns & Collection Tubes
• Collection Tubes
• Detailed Protocol
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifications
Specifications
Spin Column
Spin Column
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR
250 Preps
FFPE DNA Purification Kit
Genomic DNA
Spin Column
FFPE, Fixed Samples
Not High-throughput Compatible (Manual)
GeneJET
250 Preps
<20 min.
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.