missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ GeneJET FFPE DNA Purification Kit

Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue.

$331.00 - $1374.00

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Spin Column
Sample Type FFPE, Fixed Samples
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERK0881
View Documents
Thermo Scientific™
K0881
50 Preps
Each for $331.00
 
FERK0882
View Documents
Thermo Scientific™
K0882
250 Preps
Each for $1,374.00
 
Description

Description

Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue. Up to 8 sections (10 μm thickness) of FFPE sample from various tissues can be used for genomic DNA extraction without overnight incubation. Elimination of toxic reagents commonly used for deparaffinization (i.e., xylene) allows a safe DNA purification procedure with high DNA yields and excellent quality. Each preparation recovers up to 8 μg of genomic DNA that can be eluted in volumes from 20 μL allowing for concentrated samples. Due to high decrosslinking efficiency high quality purified DNA shows reliable performance in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or NGS library preparation.

Highlights
High quality DNA—suitable for PCR and qPCR
Fast—procedure can be completed within 2 hours
Safe—non toxic solvents
Efficient—yields up to 8 μg of purified DNA

Applications
• PCR
• qPCR
• NGS library preparation

Includes
• Proteinase K Solution
• RNase A Solution
• Digestion Buffer
• Binding Buffer
• Wash Buffer I (conc.)
• Wash Buffer II (conc.)
• Elution Buffer
• GeneJET DNA Purification Columns & Collection Tubes
• Collection Tubes
• Detailed Protocol

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

Specifications

Specifications

Spin Column
Spin Column
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR
GeneJET
Genomic DNA
Spin Column
FFPE, Fixed Samples
Not High-throughput Compatible (Manual)
FFPE DNA Purification Kit
<20 min.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.