Learn More
Thermo Scientific™ Pierce™ ECL 2 Western Blotting Substrate
Recommended HRP substrate for fluorescence imagers
$268.85 - $2,083.37
Specifications
Product Type ECL Western Blot Substrate
Kit Contents Detection reagent 1 and 2
Content And Storage 2°C to 8°C
View More Specs
Includes: Detection Reagent 1 and 2
Products 3
Catalog Number Mfr. No. Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Quantity Sufficient For Price Quantity  
PI80196
View Documents
Thermo Scientific™
80196
100 mL 10 Mini-blots/1000 cm^2 of membrane N/A
Only null left
PI80196X3
View Documents
Thermo Scientific™
LSG80196X3
3 x 100 mL 30 Mini-blots/3000 cm^2 of membrane N/A
Only null left
PI80197
View Documents
Thermo Scientific™
80197
25 mL 250 cm^2 of membrane N/A
Only null left
Description
Description
Pierce ECL 2 Substrate is a chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated antibodies and probes by Western blotting using X-ray film, CCD camera documentation systems or laser-based imagers. Pierce ECL 2 Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers.
Pierce ECL 2 substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. A broad dynamic range and excellent sensitivity combined with a long-lasting signal make Pierce ECL 2 Substrate a desirable Western blotting substrate.
Highlights:
  • High sensitivity — detect targets down to the low-picogram level
  • Long signal duration — sustained light output for as long as five hours
  • More imaging options — X-ray, CCD or laser-based imagers
  • More affordable — high quality and performance at a lower price than other suppliers' similar substrates
Specifications
Specifications
ECL Western Blot Substrate
Detection reagent 1 and 2
2°C to 8°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'