Thermo Scientific™ Septa
A wide range to cover most applications
Supplier:  Thermo Scientific™ 31303233
 View more versions of this product
Catalog No. 03-052-730
Explore available promotions
Description
Description
A wide range to cover most applications
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Specifications
Specifications
GC Instruments
Low Bleed Septum
Pack of 50
Silicone
11 mm
Product Suggestions
Product Suggestions