Thermo Scientific™ Septa
A wide range to cover most applications
$223.34 - $223.34
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity | ||||||
03-052-730
|
Thermo Scientific™
31303233 |
11 mm |
Pack of 50 Each for $223.34
|
|
||||||
Description
A wide range to cover most applications
- Variety of materials and temperature limits for all applications
- For use in Thermo Scientific TRACE™ and FOCUS™ GCs and instruments from other manufacturers
- Contaminant-free in individual blister packs