Epredia™ Ultra Disposable Microtome Blades Edge
Edge
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
Supplier:  Epredia™ 3053835
 View more versions of this product
Catalog No. 30-538-35
Edge
Explore available promotions
Description
Description
  • High- and low-profile universal blades can be used with any microtome or cryostat on the market
  • Unique angle profile that facilitates faster ribboning
  • Increased Longevity: Produces an edge that is twice as strong as traditional blades
  • Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
Specifications
Low
Microtomes, Cryostats
Single-use
34°