Epredia™ Ultra Disposable Microtome Blades
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
$459.50 - $475.12
Specifications
For Use With (Equipment) | Microtomes, Cryostats |
---|---|
Disposable | Single-use |
Cutting Angle | 34° |
Description
- High- and low-profile universal blades can be used with any microtome or cryostat on the market
- Unique angle profile that facilitates faster ribboning
- Increased Longevity: Produces an edge that is twice as strong as traditional blades
- Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
Microtomes, Cryostats | |
Single-use | |
34° |