MilliporeSigma™ Supelco™ Thermogreen™ LB-2 Septa, Solid Discs
Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
$69.92 - $3607.04
Specifications
Color | Green |
---|---|
Material | Rubber |
Style | Solid Disc |
Temperature Range (Metric) | 100°C to 350°C |
Product Line | Thermogreen |
Catalog Number | Mfr. No. | Diameter (Metric) Outer | No. per Pack | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Outer | No. per Pack | Price | Quantity | ||||
111020862
|
MilliporeSigma™ Supelco™
20638 |
5 mm | 50/Pk. |
Each for $226.65
|
|
||||
111020859
|
MilliporeSigma™ Supelco™
20651 |
6 mm | 50/Pk. |
Each for $241.51
|
|
||||
111001601
|
MilliporeSigma™ Supelco™
23157 |
10 mm | 1000/Pk. |
Each for $3,607.04
|
|
||||
111020849
|
MilliporeSigma™ Supelco™
20654 |
11 mm | 50/Pk. |
Each for $241.50
|
|
||||
111020759
|
MilliporeSigma™ Supelco™
23163 |
11 mm | 250/Pk. |
Each for $1,022.17
|
|
||||
111020695
|
MilliporeSigma™ Supelco™
23164 |
11 mm | 1000/Pk. |
Each for $3,511.98
|
|
||||
111020860
|
MilliporeSigma™ Supelco™
23154 |
11.5 mm | 50/Pk. |
Each for $223.20
|
|
||||
111016344
|
MilliporeSigma™ Supelco™
20660U |
12.5 mm | 50/Pk. |
Each for $252.25
|
|
||||
111008695
|
MilliporeSigma™ Supelco™
20678 |
12.5 mm | 250/Pk. |
Each for $952.02
|
|
||||
111010575
|
MilliporeSigma™ Supelco™
20663 |
16 mm | 50/Pk. |
Each for $255.95
|
|
||||
111020861
|
MilliporeSigma™ Supelco™
23159 |
17 mm | 50/Pk. |
Each for $223.20
|
|
||||
111011627
|
MilliporeSigma™ Supelco™
28006U |
9 mm | 5/Pk. |
Each for $69.92
|
|
||||
111020850
|
MilliporeSigma™ Supelco™
28021U |
9 mm | 50/Pk. |
Each for $235.91
|
|
||||
111020863
|
MilliporeSigma™ Supelco™
20652 |
9.5 mm | 50/Pk. |
Each for $223.20
|
|
||||
111020761
|
MilliporeSigma™ Supelco™
20666 |
9.5 mm | 250/Pk. |
Each for $961.93
|
|
||||
111020851
|
MilliporeSigma™ Supelco™
20653U |
10 mm | 50/Pk. |
Each for $244.93
|
|
||||
111020760
|
MilliporeSigma™ Supelco™
23156 |
10 mm | 250/Pk. |
Each for $958.51
|
|
||||
Description
- An improved version over the original Thermogreen™ LB-1 Septa
- Extremely low bleed over a wide range of inlet temperatures (100° to 350°C)
- Easy needle penetration
- High puncture tolerance (ideal for autosamplers)
- Rubber formulation exclusive to Supelco™
- Pre-conditioned, ready to use
Specifications
Green | |
Solid Disc | |
Thermogreen |
Rubber | |
100°C to 350°C | |
Septa, LB-2 |
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC